tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-[2-(4-Amino-1H-pyrazol-1-yl)ethyl]piperazine, N1-BOC protected

081336-1G
102834-638EA 553.85 USD
102834-638
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate ≥97%
tert-Butyl-4-[2-(4-amino-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₅N₅O₂
MW: 295.39 g/mol
MDL Number: MFCD11505005
CAS Number: 1171901-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR