4-Cyclohex-1-en-1-yl-1H-pyrazol-5-amine ≥97%

Supplier: Matrix Scientific

081332-1G
102834-630EA 806.15 USD
102834-630
4-Cyclohex-1-en-1-yl-1H-pyrazol-5-amine ≥97%
4-Cyclohex-1-en-1-yl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₉H₁₃N₃
MDL Number: MFCD09991752
CAS Number: 1257859-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR