3-Aminothien-2-yl acetate ≥97%

Supplier: Matrix Scientific

081320-1G
102834-596EA 1203.08 USD
102834-596
3-Aminothien-2-yl acetate ≥97%
3-Aminothien-2-yl acetate

Hazard: Irritant

Formula: C₆H₇NO₂S
MDL Number: MFCD08445662
CAS Number: 914636-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR