Methyl-3-[(3-Aminothien-2-yl)thio]propanoate ≥95%

Supplier: Matrix Scientific

081319-1G
102834-594EA 863.08 USD
102834-594
Methyl-3-[(3-Aminothien-2-yl)thio]propanoate ≥95%
Methyl-3-[(3-Aminothien-2-yl)thio]propanoate

Hazard: Irritant

Formula: C₈H₁₁NO₂S₂
MDL Number: MFCD16622230
CAS Number: 1287218-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR