Methyl [(3-aminothien-2-yl)thio]acetate ≥95%

Supplier: Matrix Scientific

081318-1G
102834-592EA 863.08 USD
102834-592
Methyl [(3-aminothien-2-yl)thio]acetate ≥95%
Methyl [(3-aminothien-2-yl)thio]acetate

Hazard: Irritant

Formula: C₇H₉NO₂S₂
MDL Number: MFCD16622229
CAS Number: 1287218-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR