2-Phenoxythien-3-ylamine ≥95%

Supplier: Matrix Scientific

081317-1G
102834-590EA 863.08 USD
102834-590
2-Phenoxythien-3-ylamine ≥95%
2-Phenoxythien-3-ylamine

Hazard: Irritant

Formula: C₁₀H₉NOS
MDL Number: MFCD16622228
CAS Number: 1287217-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR