2-Ethoxythien-3-ylamine ≥95%
Supplier: Matrix Scientific
081316-1G
102834-588EA
863.08
USD
102834-588
2-Ethoxythien-3-ylamine ≥95%
2-Ethoxythien-3-ylamine
Hazard: Irritant
Formula:
C₆H₉NOS |
MDL Number:
MFCD16622227 CAS Number: 1287217-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...