2-[(3-Aminopyridin-2-yl)(cyclohexyl)amino]ethanol ≥97%

Supplier: Matrix Scientific

081281-1G
102834-508EA 949.23 USD
102834-508
2-[(3-Aminopyridin-2-yl)(cyclohexyl)amino]ethanol ≥97%
2-[(3-Aminopyridin-2-yl)(cyclohexyl)amino]ethanol

Hazard: Irritant

Formula: C₁₃H₂₁N₃O
MDL Number: MFCD16262074
CAS Number: 1308158-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR