4-Thien-2-ylpyridin-2-amine ≥97%
Supplier: Matrix Scientific
081275-1G
102834-494EA
1118.46
USD
102834-494
4-Thien-2-ylpyridin-2-amine ≥97%
4-Thien-2-ylpyridin-2-amine
Hazard: Irritant
Formula:
C₉H₈N₂S |
MDL Number:
MFCD12032858 CAS Number: 1159815-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...