2-(Tetrahydro-2H-pyran-2-ylmethoxy)pyridin-3-amine ≥97%
Supplier: Matrix Scientific
081271-1G
102834-482EA
750.77
USD
102834-482
2-(Tetrahydro-2H-pyran-2-ylmethoxy)pyridin-3-amine ≥97%
2-(Tetrahydro-2H-pyran-2-ylmethoxy)pyridin-3-amine
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₂ |
MDL Number:
MFCD09812500 CAS Number: 1016684-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...