2-(Cyclobutylmethoxy)pyridin-3-amine ≥97%

Supplier: Matrix Scientific

081270-1G
102834-480EA 750.77 USD
102834-480
2-(Cyclobutylmethoxy)pyridin-3-amine ≥97%
2-(Cyclobutylmethoxy)pyridin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MDL Number: MFCD14666455
CAS Number: 1287217-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR