2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-3-amine ≥97%
Supplier: Matrix Scientific
081269-1G
102834-478EA
750.77
USD
102834-478
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-3-amine ≥97%
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-3-amine
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₂ |
MDL Number:
MFCD14607464 CAS Number: 1211758-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...