6-(Tetrahydro-2H-pyran-2-ylmethoxy)pyridin-3-amine ≥97%

Supplier: Matrix Scientific

081267-1G
102834-474EA 750.77 USD
102834-474
6-(Tetrahydro-2H-pyran-2-ylmethoxy)pyridin-3-amine ≥97%
6-(Tetrahydro-2H-pyran-2-ylmethoxy)pyridin-3-amine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD09816925
CAS Number: 1016745-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR