6-(Cyclobutylmethoxy)pyridin-3-amine ≥97%

Supplier: Matrix Scientific

081266-1G
102834-472EA 750.77 USD
102834-472
6-(Cyclobutylmethoxy)pyridin-3-amine ≥97%
6-(Cyclobutylmethoxy)pyridin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MDL Number: MFCD14666454
CAS Number: 1287218-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR