3-Amino-6-chloroisonicotinamide ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-chloroisonicotinamide

081248-1G
102834-430EA 784.62 USD
102834-430
3-Amino-6-chloroisonicotinamide ≥97%
3-Amino-6-chloroisonicotinamide

Hazard: Irritant

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD12026342
CAS Number: 1217026-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR