3-Amino-6-pyrrolidin-1-ylpyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

081242-1G
102834-414EA 750.77 USD
102834-414
3-Amino-6-pyrrolidin-1-ylpyridine-2-carbonitrile ≥97%
3-Amino-6-pyrrolidin-1-ylpyridine-2-carbonitrile

Hazard: Irritant

Formula: C₁₀H₁₂N₄
MDL Number: MFCD12026340
CAS Number: 1215553-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR