3-Amino-6-(4-methylpiperazin-1-yl)pyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

081241-1G
102834-412EA 750.77 USD
102834-412
3-Amino-6-(4-methylpiperazin-1-yl)pyridine-2-carbonitrile ≥97%
3-Amino-6-(4-methylpiperazin-1-yl)pyridine-2-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₅N₅
MDL Number: MFCD12026339
CAS Number: 1216615-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR