3-Amino-6-piperidin-1-ylpyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

081240-1G
102834-410EA 750.77 USD
102834-410
3-Amino-6-piperidin-1-ylpyridine-2-carbonitrile ≥97%
3-Amino-6-piperidin-1-ylpyridine-2-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₄N₄
MDL Number: MFCD12026338
CAS Number: 1215852-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR