3-Amino-6-morpholin-4-ylpyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

081239-1G
102834-408EA 750.77 USD
102834-408
3-Amino-6-morpholin-4-ylpyridine-2-carbonitrile ≥97%
3-Amino-6-morpholin-4-ylpyridine-2-carbonitrile

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MDL Number: MFCD12026337
CAS Number: 1215366-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR