tert-Butyl-4-[(5-aminopyridin-2-yl)oxy]piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

081235-1G
102834-400EA 613.85 USD
102834-400
tert-Butyl-4-[(5-aminopyridin-2-yl)oxy]piperidine-1-carboxylate ≥97%
tert-Butyl-4-[(5-aminopyridin-2-yl)oxy]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₃N₃O₃
MW: 293.37 g/mol
MDL Number: MFCD12026334
CAS Number: 346665-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR