6-(Cyclohexyloxy)pyridin-3-amine ≥97%
Supplier: Matrix Scientific
081234-1G
102834-398EA
553.85
USD
102834-398
6-(Cyclohexyloxy)pyridin-3-amine ≥97%
6-(Cyclohexyloxy)pyridin-3-amine
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O |
MDL Number:
MFCD09729052 CAS Number: 224187-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...