6-(3-Bromophenoxy)pyridin-3-amine ≥97%

Supplier: Matrix Scientific

081231-1G
102834-392EA 496.92 USD
102834-392
6-(3-Bromophenoxy)pyridin-3-amine ≥97%
6-(3-Bromophenoxy)pyridin-3-amine

Hazard: Irritant

Formula: C₁₁H₉BrN₂O
MDL Number: MFCD09740262
CAS Number: 954567-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR