3-{[2-(Trifluoromethyl)pyrimidin-4-yl]oxy}aniline ≥97%

Supplier: Matrix Scientific

081194-1G
102834-306EA 383.08 USD
102834-306
3-{[2-(Trifluoromethyl)pyrimidin-4-yl]oxy}aniline ≥97%
3-{[2-(Trifluoromethyl)pyrimidin-4-yl]oxy}aniline

Hazard: Irritant

Formula: C₁₁H₈F₃N₃O
MDL Number: MFCD09864894
CAS Number: 1086378-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR