5-(2-Aminophenyl)cyclohexane-1,3-dione ≥97%

Supplier: Matrix Scientific

081140-1G
102834-148EA 723.08 USD
102834-148
5-(2-Aminophenyl)cyclohexane-1,3-dione ≥97%
5-(2-Aminophenyl)cyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD12963752
CAS Number: 1215701-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR