4-(3-Bromopropoxy)aniline ≥97%

Supplier: Matrix Scientific

081126-1G
102834-120EA 553.85 USD
102834-120
4-(3-Bromopropoxy)aniline ≥97%
4-(3-Bromopropoxy)aniline

Hazard: Irritant

Formula: C₉H₁₂BrNO
MDL Number: MFCD12404886
CAS Number: 143667-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR