4-(Thien-2-ylthio)aniline ≥97%

Supplier: Matrix Scientific

081125-1G
102834-118EA 636.92 USD
102834-118
4-(Thien-2-ylthio)aniline ≥97%
4-(Thien-2-ylthio)aniline

Hazard: Irritant

Formula: C₁₀H₉NS₂
MDL Number: MFCD02089756
CAS Number: 649665-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR