1,2-Diamino-3-fluoro-5-iodobenzene ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-5-iodobenzene-1,2-diamine
081115-1G
102834-090EA
707.69
USD
102834-090
1,2-Diamino-3-fluoro-5-iodobenzene ≥97%
1,2-Diamino-3-fluoro-5-iodobenzene
Hazard: Irritant
Formula:
C₆H₆FIN₂ MW: 252.03 g/mol |
MDL Number:
MFCD03094277 CAS Number: 517920-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...