3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}aniline ≥97%

Supplier: Matrix Scientific

081109-1G
102834-074EA 524.62 USD
102834-074
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}aniline ≥97%
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}aniline

Hazard: Irritant

Formula: C₁₁H₈F₃N₃O
MDL Number: MFCD11505004
CAS Number: 1216999-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR