Methyl N-(methoxyacetyl)-2-methylalaninate
Supplier: Matrix Scientific
081051-1G
081051-500MG
102672-548EA
387.69
USD
102672-548
102672-550
Methyl N-(methoxyacetyl)-2-methylalaninate
Methyl N-(methoxyacetyl)-2-methylalaninate
Hazard: Irritant
Formula:
C₈H₁₅NO₄ |
MDL Number:
MFCD20357214 CAS Number: 1343038-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...