Methyl-N-isobutyryl-2-methylalaninate
Supplier: Matrix Scientific
081050-1G
081050-500MG
102672-546EA
484.62
USD
102672-546
102672-544
Methyl-N-isobutyryl-2-methylalaninate
Methyl-N-isobutyryl-2-methylalaninate
Hazard: Irritant
Formula:
C₉H₁₇NO₃ |
MDL Number:
MFCD20357213 CAS Number: 445396-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...