Methyl-2-methyl-N-propionylalaninate
Supplier: Matrix Scientific
081049-500MG
081049-1G
102672-542EA
484.62
USD
102672-542
102672-540
Methyl-2-methyl-N-propionylalaninate
Methyl-2-methyl-N-propionylalaninate
Hazard: Irritant
Formula:
C₈H₁₅NO₃ |
MDL Number:
MFCD20357212 CAS Number: 1343350-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...