Ethyl [1,2,4]triazolo[4,3-a]pyridine-8-carboxylate

Supplier: Matrix Scientific

081024-500MG
102672-432EA 516.92 USD
102672-432
Ethyl [1,2,4]triazolo[4,3-a]pyridine-8-carboxylate
Ethyl [1,2,4]triazolo[4,3-a]pyridine-8-carboxylate

Hazard: Irritant

Formula: C₉H₉N₃O₂
MDL Number: MFCD22124357
CAS Number: 1394306-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR