(1E)-N',2-Dihydroxy-3-methylbutanimidamide
Supplier: Matrix Scientific
081005-1G
081005-500MG
102672-338EA
516.92
USD
102672-338
102672-340
(1E)-N',2-Dihydroxy-3-methylbutanimidamide
(1E)-N',2-Dihydroxy-3-methylbutanimidamide
Hazard: Irritant
Formula:
C₅H₁₂N₂O₂ |
MDL Number:
MFCD22422038 CAS Number: 1394306-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...