4-(Azidomethyl)-N,N-dimethylbenzamide
Supplier: Matrix Scientific
081004-500MG
081004-5G
081004-1G
102672-336EA
1615.38
USD
102672-336
102672-334
102672-332
4-(Azidomethyl)-N,N-dimethylbenzamide
4-(Azidomethyl)-N,N-dimethylbenzamide
Hazard: Irritant
Formula:
C₁₀H₁₂N₄O |
MDL Number:
MFCD22422037 CAS Number: 1394306-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...