3-(Bromomethyl)-5-isopropyl-1,2,4-oxadiazole
Supplier: Matrix Scientific
081001-500MG
081001-5G
081001-1G
102672-318EA
646.15
USD
102672-318
102672-316
102672-320
3-(Bromomethyl)-5-isopropyl-1,2,4-oxadiazole
3-(Bromomethyl)-5-isopropyl-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₆H₉BrN₂O |
MDL Number:
MFCD22422035 CAS Number: 1312700-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...