(1Z)-N'-Hydroxy-2-methoxypropanimidamide

Supplier: Matrix Scientific

081000-500MG 081000-1G
102672-314EA 807.69 USD
102672-314 102672-312
(1Z)-N'-Hydroxy-2-methoxypropanimidamide
(1Z)-N'-Hydroxy-2-methoxypropanimidamide

Hazard: Irritant

Formula: C₄H₁₀N₂O₂
MDL Number: MFCD16078779
CAS Number: 1251430-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR