(2Z)-2-Amino-2-(hydroxyimino)-N,N-dimethylacetamide
Supplier: Matrix Scientific
080987-1G
080987-500MG
102672-246EA
646.15
USD
102672-246
102672-244
(2Z)-2-Amino-2-(hydroxyimino)-N,N-dimethylacetamide
(2Z)-2-Amino-2-(hydroxyimino)-N,N-dimethylacetamide
Hazard: Irritant
Formula:
C₄H₉N₃O₂ |
MDL Number:
MFCD22422031 CAS Number: 1393733-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...