4-(Chloromethyl)-5-methyl-1,3-thiazole hydrochloride
Supplier: Matrix Scientific
080983-1G
080983-500MG
102672-226EA
823.08
USD
102672-226
102672-224
4-(Chloromethyl)-5-methyl-1,3-thiazole hydrochloride
4-(Chloromethyl)-5-methyl-1,3-thiazole hydrochloride
Hazard: Irritant
Formula:
C₅H₇Cl₂NS |
MDL Number:
MFCD22046795 CAS Number: 141305-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...