(2Z)-3-(5-Bromo-1H-1,2,4-triazol-3-yl)acrylic acid

Supplier: Matrix Scientific

080971-1G 080971-500MG
102672-162EA 646.15 USD
102672-162 102672-160
(2Z)-3-(5-Bromo-1H-1,2,4-triazol-3-yl)acrylic acid
(2Z)-3-(5-Bromo-1H-1,2,4-triazol-3-yl)acrylic acid

Hazard: Irritant

Formula: C₅H₄BrN₃O₂
MDL Number: MFCD22422025
CAS Number: 1394306-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR