5-Pyrrolidin-3-ylpyrimidine dihydrochloride
Supplier: Matrix Scientific
Synonyms:
5-(Pyrrolidin-3-yl)pyrimidine dihydrochloride
080970-500MG
080970-1G
102672-158EA
807.69
USD
102672-158
102672-156
5-Pyrrolidin-3-ylpyrimidine dihydrochloride
5-Pyrrolidin-3-ylpyrimidine dihydrochloride
Hazard: Irritant
Formula:
C₈H₁₃Cl₂N₃ MW: 222.12 g/mol |
MDL Number:
MFCD22422024 CAS Number: 1417569-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...