3-(Bromomethyl)-4-methyl-4H-1,2,4-triazole hydrobromide
Supplier: Matrix Scientific
080957-500MG
080957-5G
080957-1G
102672-086EA
2038.46
USD
102672-086
102672-084
102672-082
3-(Bromomethyl)-4-methyl-4H-1,2,4-triazole hydrobromide
3-(Bromomethyl)-4-methyl-4H-1,2,4-triazole hydrobromide
Hazard: Irritant
Formula:
C₄H₇Br₂N₃ |
MDL Number:
MFCD22422019 CAS Number: 1417566-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...