[2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethyl]amine hydrochloride
Supplier: Matrix Scientific
Synonyms:
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanamine hydrochloride
080929-1G
080929-500MG
102671-942EA
646.15
USD
102671-942
102671-944
[2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethyl]amine hydrochloride
[2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethyl]amine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₂ClN₃ MW: 197.67 g/mol |
MDL Number:
MFCD22422013 CAS Number: 1417567-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...