N-(2,4-Dimethylphenyl)-1H-pyrazole-3-carboxamide

Supplier: Matrix Scientific

080917-500MG 080917-1G
102671-884EA 387.69 USD
102671-884 102671-886
N-(2,4-Dimethylphenyl)-1H-pyrazole-3-carboxamide
N-(2,4-Dimethylphenyl)-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₃N₃O
MDL Number: MFCD02128530
CAS Number: 305346-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR