N-(4-Methyl-1,3-thiazol-2-yl)-3-(4-oxoquinazolin-3(4H)-yl)propanamide

Supplier: Matrix Scientific

080916-500MG
102671-882EA 615.38 USD
102671-882
N-(4-Methyl-1,3-thiazol-2-yl)-3-(4-oxoquinazolin-3(4H)-yl)propanamide
N-(4-Methyl-1,3-thiazol-2-yl)-3-(4-oxoquinazolin-3(4H)-yl)propanamide

Hazard: Irritant

Formula: C₁₅H₁₄N₄O₂S
MDL Number: MFCD08749650
CAS Number: 919747-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR