N-(2-Hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
Supplier: Matrix Scientific
080912-500MG
080912-1G
102671-860EA
193.85
USD
102671-860
102671-862
N-(2-Hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
N-(2-Hydroxyphenyl)-5-oxo-1-phenylpyrrolidine-3-carboxamide
Hazard: Irritant
Formula:
C₁₇H₁₆N₂O₃ |
MDL Number:
MFCD04083670 CAS Number: 591213-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...