N-[2-(2,4-Dioxo-1,3-thiazolidin-3-yl)ethyl]-5-oxo-1-phenylpyrrolidine-3-carboxamide
Supplier: Matrix Scientific
080909-1G
080909-500MG
102671-850EA
243.08
USD
102671-850
102671-848
N-[2-(2,4-Dioxo-1,3-thiazolidin-3-yl)ethyl]-5-oxo-1-phenylpyrrolidine-3-carboxamide
N-[2-(2,4-Dioxo-1,3-thiazolidin-3-yl)ethyl]-5-oxo-1-phenylpyrrolidine-3-carboxamide
Hazard: Irritant
Formula:
C₁₆H₁₇N₃O₄S |
MDL Number:
MFCD02152999 CAS Number: 351066-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...