2-Methoxy-4-{(Z)-[(2-methoxyphenyl)imino]methyl}phenol hydrochloride
Supplier: Matrix Scientific
080895-1G
080895-5G
080895-500MG
102671-798EA
161.54
USD
102671-798
102671-800
102671-802
2-Methoxy-4-{(Z)-[(2-methoxyphenyl)imino]methyl}phenol hydrochloride
2-Methoxy-4-{(Z)-[(2-methoxyphenyl)imino]methyl}phenol hydrochloride
Hazard: Irritant
Formula:
C₁₅H₁₆ClNO₃ |
MDL Number:
MFCD22422010 CAS Number: 1417574-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...