(1-Cyclopropyl-2,4,6-trioxo-2,3,4,5,6,7-hexahydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)acetic acid
Supplier: Matrix Scientific
080890-5G
080890-500MG
080890-1G
102671-782EA
323.08
USD
102671-782
102671-784
102671-780
(1-Cyclopropyl-2,4,6-trioxo-2,3,4,5,6,7-hexahydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)acetic acid
(1-Cyclopropyl-2,4,6-trioxo-2,3,4,5,6,7-hexahydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)acetic acid
Hazard: Irritant
Formula:
C₁₁H₁₁N₃O₅ |
MDL Number:
MFCD22374998 CAS Number: 1379811-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...