4,6-Diamino-1-cyclopropylpyrimidin-2(1H)-one

Supplier: Matrix Scientific

080889-1G 080889-500MG 080889-5G
102671-774EA 387.69 USD
102671-774 102671-776 102671-778
4,6-Diamino-1-cyclopropylpyrimidin-2(1H)-one
4,6-Diamino-1-cyclopropylpyrimidin-2(1H)-one

Hazard: Irritant

Formula: C₇H₁₀N₄O
MDL Number: MFCD22374997
CAS Number: 1379811-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR