7-Amino-2-cyclobutyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
Supplier: Matrix Scientific
080886-1G
080886-500MG
080886-5G
102671-760EA
1615.38
USD
102671-760
102671-756
102671-758
7-Amino-2-cyclobutyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
7-Amino-2-cyclobutyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₁N₅O₂ MW: 233.23 g/mol |
MDL Number:
MFCD22374995 CAS Number: 1379811-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...